Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGGTGACGTAATGAAATTACAG[A/G]TATGGCTAGTCTTTTTTAATAGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611911 MIM: 611684 | ||||||||||||||||||||
Literature Links: |
ISCU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISCU - iron-sulfur cluster assembly enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301140.1 | Intron | NP_001288069.1 | ||||
NM_001301141.1 | Intron | NP_001288070.1 | ||||
NM_001320042.1 | Intron | NP_001306971.1 | ||||
NM_014301.4 | Intron | NP_055116.1 | ||||
NM_213595.3 | Intron | NP_998760.1 | ||||
XM_011538100.2 | Intron | XP_011536402.1 | ||||
XM_011538101.2 | Intron | XP_011536403.1 |
SART3 - squamous cell carcinoma antigen recognized by T-cells 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |