Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATGCTTAAGCAACAGGGTGCCAA[A/G]GAGAAGGGCTTGCCACCCAGCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606778 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SSH1 - slingshot protein phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161330.1 | 6585 | Intron | NP_001154802.1 | |||
NM_001161331.1 | 6585 | Intron | NP_001154803.1 | |||
NM_018984.3 | 6585 | UTR 3 | NP_061857.3 | |||
XM_005268984.4 | 6585 | UTR 3 | XP_005269041.1 | |||
XM_005268985.2 | 6585 | UTR 3 | XP_005269042.1 | |||
XM_011538497.1 | 6585 | UTR 3 | XP_011536799.1 | |||
XM_011538499.1 | 6585 | UTR 3 | XP_011536801.1 | |||
XM_011538500.2 | 6585 | UTR 3 | XP_011536802.1 | |||
XM_011538501.2 | 6585 | UTR 3 | XP_011536803.1 | |||
XM_017019491.1 | 6585 | UTR 3 | XP_016874980.1 | |||
XM_017019492.1 | 6585 | UTR 3 | XP_016874981.1 | |||
XM_017019493.1 | 6585 | UTR 3 | XP_016874982.1 | |||
XM_017019494.1 | 6585 | UTR 3 | XP_016874983.1 |