Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCACTGGACATAAAGGTCTACG[C/T]GGACCGCTTCACCCCACTCACCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAO - D-amino acid oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001917.4 | 350 | Missense Mutation | GCG,GTG | A,V 36 | NP_001908.3 | |
XM_005268692.3 | 350 | Missense Mutation | GCG,GTG | A,V 36 | XP_005268749.1 | |
XM_011538004.2 | 350 | Missense Mutation | GCG,GTG | A,V 36 | XP_011536306.1 | |
XM_011538005.2 | 350 | Missense Mutation | GCG,GTG | A,V 36 | XP_011536307.1 |