Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAGCTCTACATCAGCATCTGAAA[A/G]GTTGTGGAAATTGTGTCCATGAAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603104 | |||||||||||||||||||||||
Literature Links: |
CPN2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CPN2 - carboxypeptidase N subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC15 - leucine rich repeat containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135057.2 | 5335 | UTR 3 | NP_001128529.2 | |||
NM_130830.4 | 5335 | UTR 3 | NP_570843.2 |