Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCACGGGGAGCGCCTAGTCCTCC[A/G]GGATGGGAGTGTTGCAGTTCCCGTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614552 | |||||||||||||||||||||||
Literature Links: |
XXYLT1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
XXYLT1 - xyloside xylosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308069.1 | 1280 | Missense Mutation | CCG,CTG | P,L 245 | NP_001294998.1 | |
NM_152531.4 | 1280 | Missense Mutation | CCG,CTG | P,L 391 | NP_689744.3 | |
XM_005269286.4 | 1280 | Missense Mutation | CCG,CTG | P,L 204 | XP_005269343.1 | |
XM_011512448.2 | 1280 | Missense Mutation | CCG,CTG | P,L 204 | XP_011510750.1 | |
XM_017005749.1 | 1280 | Intron | XP_016861238.1 | |||
XM_017005750.1 | 1280 | Missense Mutation | CCG,CTG | P,L 188 | XP_016861239.1 |