Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAATTTAAAGCCTTGCTGTTTAC[C/T]CCATCATTTCTGCCAAATGTAGTCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607766 MIM: 614552 | |||||||||||||||||||||||
Literature Links: |
ACAP2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACAP2 - ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012287.5 | 5088 | UTR 3 | NP_036419.3 | |||
XM_006713557.3 | 5088 | UTR 3 | XP_006713620.1 | |||
XM_011512602.2 | 5088 | Intron | XP_011510904.1 | |||
XM_011512603.2 | 5088 | Intron | XP_011510905.1 | |||
XM_011512604.2 | 5088 | Intron | XP_011510906.1 | |||
XM_011512605.2 | 5088 | Intron | XP_011510907.1 | |||
XM_011512606.2 | 5088 | Intron | XP_011510908.1 | |||
XM_017006046.1 | 5088 | UTR 3 | XP_016861535.1 | |||
XM_017006047.1 | 5088 | Intron | XP_016861536.1 | |||
XM_017006048.1 | 5088 | UTR 3 | XP_016861537.1 | |||
XM_017006049.1 | 5088 | UTR 3 | XP_016861538.1 |
XXYLT1 - xyloside xylosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |