Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAACCAAAATTTTATGGCTCAA[C/T]AGTCTAACCACTTAAAGTTCTGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601448 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPAT - nuclear protein, coactivator of histone transcription | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321307.1 | 5887 | UTR 3 | NP_001308236.1 | |||
NM_002519.2 | 5887 | UTR 3 | NP_002510.2 | |||
XM_011542854.2 | 5887 | UTR 3 | XP_011541156.1 | |||
XM_017017802.1 | 5887 | UTR 3 | XP_016873291.1 | |||
XM_017017803.1 | 5887 | UTR 3 | XP_016873292.1 |