Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTACTTACTGAGCTGTGCCTCT[C/G]TATATCCTGTAACATTTCTGTGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601448 | ||||||||||||||||||||
Literature Links: |
NPAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPAT - nuclear protein, coactivator of histone transcription | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321307.1 | 4618 | Missense Mutation | CAG,GAG | Q,E 1258 | NP_001308236.1 | |
NM_002519.2 | 4618 | Missense Mutation | CAG,GAG | Q,E 1251 | NP_002510.2 | |
XM_011542854.2 | 4618 | Missense Mutation | CAG,GAG | Q,E 1260 | XP_011541156.1 | |
XM_017017802.1 | 4618 | Missense Mutation | CAG,GAG | Q,E 1192 | XP_016873291.1 | |
XM_017017803.1 | 4618 | Missense Mutation | CAG,GAG | Q,E 1192 | XP_016873292.1 |