Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAAGTCTTAATATGTAGGACTG[A/G]TTAGTTATAAAGTGAAATCGACTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP20 - Rho GTPase activating protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258415.1 | 5398 | UTR 3 | NP_001245344.1 | |||
NM_001258416.1 | 5398 | UTR 3 | NP_001245345.1 | |||
NM_001258417.1 | 5398 | UTR 3 | NP_001245346.1 | |||
NM_001258418.1 | 5398 | UTR 3 | NP_001245347.1 | |||
NM_020809.3 | 5398 | UTR 3 | NP_065860.2 | |||
XM_005271628.3 | 5398 | UTR 3 | XP_005271685.1 | |||
XM_006718893.3 | 5398 | UTR 3 | XP_006718956.1 |