Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCAGGGAGATGTGTTCTTTTTC[A/G]GAAACATCTTTGTGTTTAGCAGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610493 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf52 - chromosome 11 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DIXDC1 - DIX domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037954.3 | Intron | NP_001033043.1 | ||||
NM_001278542.1 | Intron | NP_001265471.1 | ||||
NM_033425.4 | Intron | NP_219493.1 | ||||
XM_017018466.1 | Intron | XP_016873955.1 | ||||
XM_017018467.1 | Intron | XP_016873956.1 | ||||
XM_017018468.1 | Intron | XP_016873957.1 | ||||
XM_017018469.1 | Intron | XP_016873958.1 |
HSPB2-C11orf52 - HSPB2-C11orf52 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |