Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGACGGAGTACGCCATCAAGTG[C/G]GCCCCCTGCCTTCCACCCGACGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608774 MIM: 610732 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKK1 - ankyrin repeat and kinase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178510.1 | 2027 | Missense Mutation | TGC,TGG | C,W 52 | NP_848605.1 | |
XM_011542736.2 | 2027 | Missense Mutation | TGC,TGG | C,W 52 | XP_011541038.1 | |
XM_011542737.2 | 2027 | Missense Mutation | TGC,TGG | C,W 52 | XP_011541039.1 | |
XM_011542738.2 | 2027 | Intron | XP_011541040.1 | |||
XM_017017475.1 | 2027 | Missense Mutation | TGC,TGG | C,W 52 | XP_016872964.1 |
TTC12 - tetratricopeptide repeat domain 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |