Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGTTAGCAGCCCAGAGAAAAAG[C/T]CCACTAGATTTGTCATGGCTACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604654 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTR3B - 5-hydroxytryptamine receptor 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006028.4 | Intron | NP_006019.1 | ||||
XM_011543063.1 | Intron | XP_011541365.1 | ||||
XM_017018552.1 | Intron | XP_016874041.1 |