Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATAACAATATTTTTGCCACTGG[A/G]TGTTGATGTTCCTATCCAAATCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NXPE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NXPE2 - neurexophilin and PC-esterase domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182495.5 | 895 | Intron | NP_872301.2 | |||
XM_011542604.2 | 895 | Intron | XP_011540906.1 | |||
XM_017017206.1 | 895 | Intron | XP_016872695.1 | |||
XM_017017207.1 | 895 | Intron | XP_016872696.1 | |||
XM_017017208.1 | 895 | Intron | XP_016872697.1 | |||
XM_017017209.1 | 895 | Intron | XP_016872698.1 | |||
XM_017017210.1 | 895 | Intron | XP_016872699.1 | |||
XM_017017211.1 | 895 | Intron | XP_016872700.1 | |||
XM_017017212.1 | 895 | Intron | XP_016872701.1 |
NXPE4 - neurexophilin and PC-esterase domain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077639.1 | 895 | Missense Mutation | ACC,ATC | T,I 393 | NP_001071107.1 | |
NM_017678.2 | 895 | Missense Mutation | ACC,ATC | T,I 109 | NP_060148.2 | |
XM_006718865.3 | 895 | Intron | XP_006718928.1 | |||
XM_011542881.2 | 895 | Missense Mutation | ACC,ATC | T,I 393 | XP_011541183.1 | |
XM_011542882.2 | 895 | Missense Mutation | ACC,ATC | T,I 324 | XP_011541184.1 | |
XM_011542883.2 | 895 | Intron | XP_011541185.1 |