Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACGTGACTCCTCTACCTTTTCCC[A/G]AGGCTCCCCATCTAAGATATGTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CADM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CADM1 - cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098517.1 | 2894 | UTR 3 | NP_001091987.1 | |||
NM_001301043.1 | 2894 | UTR 3 | NP_001287972.1 | |||
NM_001301044.1 | 2894 | UTR 3 | NP_001287973.1 | |||
NM_001301045.1 | 2894 | UTR 3 | NP_001287974.1 | |||
NM_014333.3 | 2894 | UTR 3 | NP_055148.3 | |||
XM_005271494.2 | 2894 | UTR 3 | XP_005271551.1 | |||
XM_017017457.1 | 2894 | UTR 3 | XP_016872946.1 | |||
XM_017017458.1 | 2894 | UTR 3 | XP_016872947.1 | |||
XM_017017459.1 | 2894 | UTR 3 | XP_016872948.1 | |||
XM_017017460.1 | 2894 | UTR 3 | XP_016872949.1 | |||
XM_017017461.1 | 2894 | UTR 3 | XP_016872950.1 |