Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGCGAGTGAAGGCAGCTATCTG[C/T]AGGTAGGTGTCCTGGCGGAAAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606368 MIM: 603901 | ||||||||||||||||||||
Literature Links: |
APOA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOA5 - apolipoprotein A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166598.1 | 955 | Silent Mutation | CTA,CTG | L,L 294 | NP_001160070.1 | |
NM_052968.4 | 955 | Silent Mutation | CTA,CTG | L,L 294 | NP_443200.2 | |
XM_017017169.1 | 955 | Silent Mutation | CTA,CTG | L,L 294 | XP_016872658.1 |
ZPR1 - ZPR1 zinc finger | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |