Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGGACACAAAGCCTCGCCATACA[C/T]GCCTCTTCCTCCTCCTGATCAGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609770 MIM: 611707 | ||||||||||||||||||||
Literature Links: |
JAML PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JAML - junction adhesion molecule like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPZL3 - myelin protein zero like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286152.1 | 680 | Silent Mutation | GCA,GCG | A,A 202 | NP_001273081.1 | |
NM_198275.2 | 680 | Silent Mutation | GCA,GCG | A,A 214 | NP_938016.1 | |
XM_006718775.2 | 680 | Silent Mutation | GCA,GCG | A,A 196 | XP_006718838.1 | |
XM_017017304.1 | 680 | Silent Mutation | GCA,GCG | A,A 198 | XP_016872793.1 |