Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATGGAGGAGCACTGTGCTGGCT[C/T]CAGAGGGGTGTTGCTTGTCTAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604873 MIM: 611707 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPZL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPZL2 - myelin protein zero like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005797.3 | 1184 | UTR 3 | NP_005788.1 | |||
NM_144765.2 | 1184 | Intron | NP_658911.1 |
MPZL3 - myelin protein zero like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |