Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCGCTCTTACGAGACGGGGGTAG[C/T]GTGCTGGGAGGAGGGAGGGAGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165360 MIM: 155735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBL - Cbl proto-oncogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCAM - melanoma cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006500.2 | 1732 | Silent Mutation | ACA,ACG | T,T 599 | NP_006491.2 | |
XM_017017759.1 | 1732 | Silent Mutation | ACA,ACG | T,T 578 | XP_016873248.1 | |
XM_017017760.1 | 1732 | Silent Mutation | ACA,ACG | T,T 567 | XP_016873249.1 | |
XM_017017761.1 | 1732 | Intron | XP_016873250.1 | |||
XM_017017762.1 | 1732 | Intron | XP_016873251.1 |
MIR6756 - microRNA 6756 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |