Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGATCTCCAGATTGGAATCCTAA[C/T]TGGAGTCCATTCCCATAGTTAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610658 | ||||||||||||||||||||
Literature Links: |
TRIM29 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM29 - tripartite motif containing 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012101.3 | Intron | NP_036233.2 | ||||
XM_005271490.1 | Intron | XP_005271547.1 | ||||
XM_011542729.1 | Intron | XP_011541031.1 | ||||
XM_011542732.1 | Intron | XP_011541034.1 | ||||
XM_011542733.1 | Intron | XP_011541035.1 | ||||
XM_017017453.1 | Intron | XP_016872942.1 | ||||
XM_017017454.1 | Intron | XP_016872943.1 |