Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAGGGGAGGGGAGAAGTGGATA[A/G]TGATTTAAACAAACAACAACAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POU2F3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POU2F3 - POU class 2 homeobox 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM136 - transmembrane protein 136 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198670.1 | 763 | Intron | NP_001185599.1 | |||
NM_001198671.1 | 763 | Intron | NP_001185600.1 | |||
NM_001198672.1 | 763 | Intron | NP_001185601.1 | |||
NM_001198673.1 | 763 | Intron | NP_001185602.1 | |||
NM_001198674.1 | 763 | Intron | NP_001185603.1 | |||
NM_001198675.1 | 763 | Intron | NP_001185604.1 | |||
NM_174926.2 | 763 | Intron | NP_777586.1 | |||
XM_017017327.1 | 763 | UTR 5 | XP_016872816.1 |