Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTGTGTGCACGGACAATGCTA[C/T]GAGGCATTGCAGTGCATTCTGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM225 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM225 - transmembrane protein 225 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013743.2 | 941 | Missense Mutation | CAT,CGT | H,R 201 | NP_001013765.2 | |
XM_006718832.3 | 941 | Missense Mutation | CAT,CGT | H,R 151 | XP_006718895.1 | |
XM_011542802.2 | 941 | Missense Mutation | CAT,CGT | H,R 156 | XP_011541104.1 | |
XM_017017653.1 | 941 | Intron | XP_016873142.1 |