Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGAGGCTTTCTTCTCGTAAGGA[A/G]TGACTGTGTTCAGATACTGCAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604825 | ||||||||||||||||||||
Literature Links: |
FEZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FEZ1 - fasciculation and elongation protein zeta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005103.4 | 1317 | Missense Mutation | ACT,ATT | T,I 346 | NP_005094.1 | |
NM_022549.3 | 1317 | Intron | NP_072043.1 | |||
XM_005271734.2 | 1317 | Missense Mutation | ACT,ATT | T,I 346 | XP_005271791.1 | |
XM_005271735.2 | 1317 | Missense Mutation | ACT,ATT | T,I 346 | XP_005271792.1 |