Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTTCACCGGAGCCATGTTCAC[C/T]TGAAGGCTGTTCATCTGAAGGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102525 MIM: 603078 | ||||||||||||||||||||
Literature Links: |
ACRV1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACRV1 - acrosomal vesicle protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001612.5 | 554 | Missense Mutation | AGT,GGT | S,G 146 | NP_001603.1 | |
NM_020069.4 | 554 | Missense Mutation | AGT,GGT | S,G 146 | NP_064454.1 | |
NM_020107.4 | 554 | Missense Mutation | AGT,GGT | S,G 91 | NP_064492.1 | |
NM_020108.4 | 554 | Missense Mutation | AGT,GGT | S,G 76 | NP_064493.1 |
CHEK1 - checkpoint kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |