Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCATAACTACATTCACGGACCAA[C/T]CAAAGCCATCTCCAAGCCTAGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM118B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM118B - family with sequence similarity 118 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPUSD4 - RNA pseudouridylate synthase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144827.1 | 2138 | UTR 3 | NP_001138299.1 | |||
NM_032795.2 | 2138 | UTR 3 | NP_116184.2 | |||
XM_011543039.2 | 2138 | UTR 3 | XP_011541341.1 |