Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTATAAACAGATTTATAGCCCT[C/T]TAAGCCATCCAATACAGATTGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600359 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNJ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNJ1 - potassium voltage-gated channel subfamily J member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000220.4 | 2180 | UTR 3 | NP_000211.1 | |||
NM_153764.2 | 2180 | UTR 3 | NP_722448.1 | |||
NM_153765.2 | 2180 | UTR 3 | NP_722449.3 | |||
NM_153766.2 | 2180 | UTR 3 | NP_722450.1 | |||
NM_153767.3 | 2180 | UTR 3 | NP_722451.1 |