Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGTATGCGAGGTTAAAAAGAC[A/G]TGGTATCAGCACTGCTTAAGGTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFRKB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFRKB - nuclear factor related to kappaB binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143835.1 | Intron | NP_001137307.1 | ||||
NM_006165.3 | Intron | NP_006156.2 | ||||
XM_005271567.3 | Intron | XP_005271624.1 | ||||
XM_005271568.3 | Intron | XP_005271625.1 | ||||
XM_011542850.2 | Intron | XP_011541152.1 | ||||
XM_011542851.2 | Intron | XP_011541153.1 | ||||
XM_011542852.2 | Intron | XP_011541154.1 | ||||
XM_017017793.1 | Intron | XP_016873282.1 | ||||
XM_017017794.1 | Intron | XP_016873283.1 | ||||
XM_017017795.1 | Intron | XP_016873284.1 | ||||
XM_017017796.1 | Intron | XP_016873285.1 | ||||
XM_017017797.1 | Intron | XP_016873286.1 | ||||
XM_017017798.1 | Intron | XP_016873287.1 | ||||
XM_017017799.1 | Intron | XP_016873288.1 | ||||
XM_017017800.1 | Intron | XP_016873289.1 | ||||
XM_017017801.1 | Intron | XP_016873290.1 |
TMEM45B - transmembrane protein 45B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |