Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAAATTGACTGAATTGATGCAG[A/C]AAAAGATTCTGAAACCCAGCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPCML PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OPCML - opioid binding protein/cell adhesion molecule like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012393.2 | 4787 | UTR 3 | NP_001012393.1 | |||
NM_001319103.1 | 4787 | UTR 3 | NP_001306032.1 | |||
NM_001319104.1 | 4787 | UTR 3 | NP_001306033.1 | |||
NM_001319105.1 | 4787 | UTR 3 | NP_001306034.1 | |||
NM_001319106.1 | 4787 | UTR 3 | NP_001306035.1 | |||
NM_002545.4 | 4787 | UTR 3 | NP_002536.1 | |||
XM_006718846.2 | 4787 | UTR 3 | XP_006718909.1 | |||
XM_011542856.2 | 4787 | UTR 3 | XP_011541158.1 |