Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCCAGCCTCTCAGGCGCAGCAG[C/T]GTTCCCGGGTGCAGGTGGAAGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF9B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF9B - immunoglobulin superfamily member 9B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277285.1 | 2523 | Missense Mutation | ACT,GCT | T,A 1337 | NP_001264214.1 | |
XM_006718795.3 | 2523 | Missense Mutation | ACT,GCT | T,A 813 | XP_006718858.1 | |
XM_011542691.2 | 2523 | Missense Mutation | ACT,GCT | T,A 1439 | XP_011540993.1 | |
XM_011542692.2 | 2523 | Missense Mutation | ACT,GCT | T,A 1435 | XP_011540994.1 | |
XM_011542694.2 | 2523 | Missense Mutation | ACT,GCT | T,A 1439 | XP_011540996.1 | |
XM_011542695.2 | 2523 | Missense Mutation | ACT,GCT | T,A 1341 | XP_011540997.1 | |
XM_017017393.1 | 2523 | Missense Mutation | ACT,GCT | T,A 1408 | XP_016872882.1 | |
XM_017017394.1 | 2523 | Intron | XP_016872883.1 | |||
XM_017017395.1 | 2523 | UTR 3 | XP_016872884.1 | |||
XM_017017396.1 | 2523 | Missense Mutation | ACT,GCT | T,A 1040 | XP_016872885.1 |