Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCGTACCGCAGGCCACCCACG[A/G]AGGCGACGGGCCACACGGACACCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151290 | ||||||||||||||||||||
Literature Links: |
B3GAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B3GAT1 - beta-1,3-glucuronyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018644.3 | 1189 | Missense Mutation | TCC,TTC | S,F 221 | NP_061114.2 | |
NM_054025.2 | 1189 | Missense Mutation | TCC,TTC | S,F 221 | NP_473366.1 | |
XM_005271506.3 | 1189 | Missense Mutation | TCC,TTC | S,F 234 | XP_005271563.1 | |
XM_011542751.2 | 1189 | Missense Mutation | TCC,TTC | S,F 234 | XP_011541053.1 | |
XM_011542753.2 | 1189 | Missense Mutation | TCC,TTC | S,F 234 | XP_011541055.1 | |
XM_017017550.1 | 1189 | Missense Mutation | TCC,TTC | S,F 221 | XP_016873039.1 | |
XM_017017551.1 | 1189 | Missense Mutation | TCC,TTC | S,F 234 | XP_016873040.1 | |
XM_017017552.1 | 1189 | Missense Mutation | TCC,TTC | S,F 117 | XP_016873041.1 |
GLB1L2 - galactosidase beta 1 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |