Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTCAATTTTTCCTGCAGTTGAA[C/T]CAGCTATTAGCTTTCTGGAGAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120353 MIM: 185260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMP1 - matrix metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145938.1 | 1532 | Nonsense Mutation | TGA,TGG | *,W 397 | NP_001139410.1 | |
NM_002421.3 | 1532 | Nonsense Mutation | TGA,TGG | *,W 463 | NP_002412.1 |
MMP10 - matrix metallopeptidase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WTAPP1 - Wilms tumor 1 associated protein pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |