Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATCATCACAGAAAATTGATATG[G/T]TACTAATTTAAATCCTGAAATGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KBTBD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KBTBD3 - kelch repeat and BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152433.3 | 3239 | UTR 3 | NP_689646.2 | |||
NM_198439.2 | 3239 | UTR 3 | NP_940841.1 | |||
XM_006718767.3 | 3239 | UTR 3 | XP_006718830.1 | |||
XM_011542618.2 | 3239 | UTR 3 | XP_011540920.1 | |||
XM_011542619.2 | 3239 | UTR 3 | XP_011540921.1 |