Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCCTCTTCTGCATGAGAGCTGT[C/T]CTCAGGGGAAATTGTAATGTATCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF479 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF479 - zinc finger protein 479 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033273.2 | Intron | NP_150376.1 | ||||
XM_011515604.2 | Intron | XP_011513906.1 | ||||
XM_011515608.2 | Intron | XP_011513910.1 | ||||
XM_011515609.2 | Intron | XP_011513911.1 | ||||
XM_011515610.2 | Intron | XP_011513912.1 | ||||
XM_011515611.2 | Intron | XP_011513913.1 | ||||
XM_017012777.1 | Intron | XP_016868266.1 | ||||
XM_017012778.1 | Intron | XP_016868267.1 |