Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTCTTACACAATGATGATCCAG[C/T]TACAAGTCAGTTTCTTCTTTTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613365 | ||||||||||||||||||||
Literature Links: |
SLCO6A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLCO6A1 - solute carrier organic anion transporter family member 6A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289002.1 | 1422 | Silent Mutation | TAA,TAG | *,* 720 | NP_001275931.1 | |
NM_001289004.1 | 1422 | Silent Mutation | TAA,TAG | *,* 658 | NP_001275933.1 | |
NM_001308014.1 | 1422 | Silent Mutation | TAA,TAG | *,* 467 | NP_001294943.1 | |
NM_173488.4 | 1422 | Silent Mutation | TAA,TAG | *,* 720 | NP_775759.3 | |
XM_005271874.3 | 1422 | Silent Mutation | TAA,TAG | *,* 720 | XP_005271931.1 | |
XM_011543147.2 | 1422 | Silent Mutation | TAA,TAG | *,* 685 | XP_011541449.1 | |
XM_011543148.2 | 1422 | Silent Mutation | TAA,TAG | *,* 641 | XP_011541450.1 | |
XM_011543150.1 | 1422 | Silent Mutation | TAA,TAG | *,* 477 | XP_011541452.1 | |
XM_011543153.2 | 1422 | Silent Mutation | TAA,TAG | *,* 446 | XP_011541455.1 | |
XM_011543154.2 | 1422 | Intron | XP_011541456.1 |