Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGGCCGAGCAGCGTCTCCCC[G/T]CGGCAGCCGGCTCCTGGCGGTGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616530 | ||||||||||||||||||||
Literature Links: |
YTHDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YTHDC2 - YTH domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022828.3 | 240 | Silent Mutation | CCG,CCT | P,P 9 | NP_073739.3 | |
XM_011543581.1 | 240 | Silent Mutation | CCG,CCT | P,P 9 | XP_011541883.1 | |
XM_011543582.2 | 240 | Silent Mutation | CCG,CCT | P,P 9 | XP_011541884.1 | |
XM_011543586.2 | 240 | Silent Mutation | CCG,CCT | P,P 9 | XP_011541888.1 | |
XM_017009730.1 | 240 | Silent Mutation | CCG,CCT | P,P 9 | XP_016865219.1 | |
XM_017009731.1 | 240 | UTR 5 | XP_016865220.1 | |||
XM_017009732.1 | 240 | UTR 5 | XP_016865221.1 | |||
XM_017009733.1 | 240 | UTR 5 | XP_016865222.1 | |||
XM_017009734.1 | 240 | UTR 5 | XP_016865223.1 | |||
XM_017009735.1 | 240 | UTR 5 | XP_016865224.1 |