Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTTTGCGTTTGTTCTTTGGCAT[C/T]GGTTTAGGATATAATCCATTTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DTWD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DTWD2 - DTW domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308081.1 | 2321 | Silent Mutation | CCA,CCG | P,P 212 | NP_001295010.1 | |
NM_173666.3 | 2321 | Silent Mutation | CCA,CCG | P,P 278 | NP_775937.1 | |
XM_011543338.2 | 2321 | Silent Mutation | CCA,CCG | P,P 434 | XP_011541640.2 | |
XM_011543340.2 | 2321 | Silent Mutation | CCA,CCG | P,P 232 | XP_011541642.1 |