Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCATGGGACATATTATGAATA[C/T]CTTTGCTTCCCAGATACATTTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107323 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH7A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH7A1 - aldehyde dehydrogenase 7 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001182.4 | 3258 | UTR 3 | NP_001173.2 | |||
NM_001201377.1 | 3258 | UTR 3 | NP_001188306.1 | |||
NM_001202404.1 | 3258 | UTR 3 | NP_001189333.2 | |||
XM_011543417.2 | 3258 | Intron | XP_011541719.1 | |||
XM_017009493.1 | 3258 | Intron | XP_016864982.1 |