Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATATATTACTGGAATTGAAGTG[A/T]CCCAGTGAAGCGTGGTGGGGAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARCH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARCH3 - membrane associated ring-CH-type finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178450.4 | 2937 | UTR 3 | NP_848545.1 | |||
XM_011543128.2 | 2937 | UTR 3 | XP_011541430.1 | |||
XM_011543129.1 | 2937 | UTR 3 | XP_011541431.1 | |||
XM_011543130.1 | 2937 | UTR 3 | XP_011541432.1 | |||
XM_011543131.2 | 2937 | UTR 3 | XP_011541433.1 | |||
XM_011543132.1 | 2937 | Intron | XP_011541434.1 | |||
XM_017009014.1 | 2937 | UTR 3 | XP_016864503.1 | |||
XM_017009015.1 | 2937 | Intron | XP_016864504.1 |