Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCAGCAACTGAGAAGTCTTCAG[C/T]CATCCTCCTGCACAGACAACTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNI2 - cyclin I family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039780.3 | 935 | Missense Mutation | CCA,TCA | P,S 357 | NP_001034869.1 | |
NM_001287252.1 | 935 | Missense Mutation | CCA,TCA | P,S 373 | NP_001274181.1 | |
NM_001287253.1 | 935 | Missense Mutation | CCA,TCA | P,S 358 | NP_001274182.1 | |
XM_017009725.1 | 935 | Missense Mutation | CCA,TCA | P,S 281 | XP_016865214.1 |
SEPT8 - septin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098811.1 | 935 | Intron | NP_001092281.1 | |||
NM_001098812.1 | 935 | Intron | NP_001092282.1 | |||
NM_001098813.1 | 935 | Intron | NP_001092283.1 | |||
NM_001300798.1 | 935 | Intron | NP_001287727.1 | |||
NM_001300799.1 | 935 | Intron | NP_001287728.1 | |||
NM_015146.1 | 935 | Intron | NP_055961.1 | |||
XM_005271927.2 | 935 | Intron | XP_005271984.1 | |||
XM_005271928.2 | 935 | Intron | XP_005271985.1 | |||
XM_005271931.3 | 935 | Intron | XP_005271988.1 | |||
XM_011543288.1 | 935 | Intron | XP_011541590.1 | |||
XM_017009254.1 | 935 | Intron | XP_016864743.1 | |||
XM_017009255.1 | 935 | Intron | XP_016864744.1 | |||
XM_017009256.1 | 935 | Intron | XP_016864745.1 | |||
XM_017009257.1 | 935 | Intron | XP_016864746.1 | |||
XM_017009258.1 | 935 | Intron | XP_016864747.1 | |||
XM_017009259.1 | 935 | Intron | XP_016864748.1 | |||
XM_017009260.1 | 935 | Intron | XP_016864749.1 | |||
XM_017009261.1 | 935 | Intron | XP_016864750.1 |