Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATTGATCTCATGAGATCTGGGG[C/T]GGGGGATGCAGGTGGCTTGGCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSTL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSTL4 - follistatin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015082.1 | 3790 | UTR 3 | NP_055897.1 | |||
XM_011543283.1 | 3790 | UTR 3 | XP_011541585.1 | |||
XM_011543284.1 | 3790 | UTR 3 | XP_011541586.1 | |||
XM_011543286.2 | 3790 | UTR 3 | XP_011541588.1 | |||
XM_011543287.2 | 3790 | UTR 3 | XP_011541589.1 | |||
XM_017009251.1 | 3790 | UTR 3 | XP_016864740.1 |