Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGACAGTAACTTCACAAATAATA[G/T]TCTATCTCCCCCAGGTGGGTAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602848 MIM: 603575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRD8 - bromodomain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164326.1 | Intron | NP_001157798.1 | ||||
NM_001300961.1 | Intron | NP_001287890.1 | ||||
NM_001300962.1 | Intron | NP_001287891.1 | ||||
NM_001300966.1 | Intron | NP_001287895.1 | ||||
NM_006696.3 | Intron | NP_006687.3 | ||||
NM_139199.1 | Intron | NP_631938.1 | ||||
XM_005271855.2 | Intron | XP_005271912.1 | ||||
XM_005271856.2 | Intron | XP_005271913.1 | ||||
XM_005271857.2 | Intron | XP_005271914.1 | ||||
XM_005271859.3 | Intron | XP_005271916.1 | ||||
XM_005271860.3 | Intron | XP_005271917.1 | ||||
XM_005271861.3 | Intron | XP_005271918.1 | ||||
XM_011543109.1 | Intron | XP_011541411.1 | ||||
XM_011543110.1 | Intron | XP_011541412.1 | ||||
XM_017008969.1 | Intron | XP_016864458.1 | ||||
XM_017008970.1 | Intron | XP_016864459.1 | ||||
XM_017008971.1 | Intron | XP_016864460.1 | ||||
XM_017008972.1 | Intron | XP_016864461.1 | ||||
XM_017008973.1 | Intron | XP_016864462.1 | ||||
XM_017008974.1 | Intron | XP_016864463.1 | ||||
XM_017008975.1 | Intron | XP_016864464.1 | ||||
XM_017008976.1 | Intron | XP_016864465.1 |
NME5 - NME/NM23 family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |