Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCGATAAGCTCATGTTCCTGTCC[A/G]GTCTACAGGGATGACCATGAGATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600285 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETF1 - eukaryotic translation termination factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256302.1 | 1507 | Silent Mutation | ACC,ACT | T,T 329 | NP_001243231.1 | |
NM_001282185.1 | 1507 | Silent Mutation | ACC,ACT | T,T 348 | NP_001269114.1 | |
NM_001291974.1 | 1507 | Silent Mutation | ACC,ACT | T,T 329 | NP_001278903.1 | |
NM_001291975.1 | 1507 | Silent Mutation | ACC,ACT | T,T 329 | NP_001278904.1 | |
NM_004730.3 | 1507 | Silent Mutation | ACC,ACT | T,T 362 | NP_004721.1 | |
XM_005271921.1 | 1507 | Silent Mutation | ACC,ACT | T,T 329 | XP_005271978.1 | |
XM_017009221.1 | 1507 | Silent Mutation | ACC,ACT | T,T 310 | XP_016864710.1 | |
XM_017009222.1 | 1507 | Silent Mutation | ACC,ACT | T,T 147 | XP_016864711.1 |