Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGAGGCCCTCGCGTCTTGCTGA[A/G]CCCGGGGAGTTAGGATGACGCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF3C5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF3C5 - general transcription factor IIIC subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122823.1 | 192 | UTR 5 | NP_001116295.1 | |||
NM_001286709.1 | 192 | UTR 5 | NP_001273638.1 | |||
NM_012087.3 | 192 | UTR 5 | NP_036219.2 | |||
XM_005272234.3 | 192 | UTR 5 | XP_005272291.1 | |||
XM_011519201.1 | 192 | UTR 5 | XP_011517503.1 | |||
XM_017015315.1 | 192 | UTR 5 | XP_016870804.1 | |||
XM_017015316.1 | 192 | UTR 5 | XP_016870805.1 |
LOC105376306 - uncharacterized LOC105376306 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |