Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCGGAATCTGCAGCCAGCATGC[C/T]CTCTGCCCGATGTCTGCAGCCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189980 MIM: 613357 MIM: 609795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABL1 - ABL proto-oncogene 1, non-receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FIBCD1 - fibrinogen C domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QRFP - pyroglutamylated RFamide peptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198180.2 | 760 | Missense Mutation | GAG,GGG | E,G 80 | NP_937823.1 |