Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGATCTCCCCGCGCCGGATCCC[A/G]AGGTGCTTGCCACCCCCGCGACGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160994 MIM: 606466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPM - heterogeneous nuclear ribonucleoprotein M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRAM1 - PML-RARA regulated adaptor molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032152.4 | 1893 | Silent Mutation | CTC,CTT | L,L 608 | NP_115528.4 | |
XM_005272502.2 | 1893 | Silent Mutation | CTC,CTT | L,L 608 | XP_005272559.1 | |
XM_011528352.2 | 1893 | Silent Mutation | CTC,CTT | L,L 610 | XP_011526654.1 | |
XM_011528353.2 | 1893 | Silent Mutation | CTC,CTT | L,L 610 | XP_011526655.1 |