Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCAGCCGCACCTCACCCCAGC[A/G]ATCTAAGAGCCCTTCACAGCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616432 MIM: 607583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF18 - Rho/Rac guanine nucleotide exchange factor 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100128573 - uncharacterized LOC100128573 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX11G - peroxisomal biogenesis factor 11 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270539.1 | Intron | NP_001257468.1 | ||||
NM_001300881.1 | Intron | NP_001287810.1 | ||||
NM_080662.3 | Intron | NP_542393.1 | ||||
XM_005272506.2 | Intron | XP_005272563.1 | ||||
XM_011528426.2 | Intron | XP_011526728.1 | ||||
XM_011528427.2 | Intron | XP_011526729.1 | ||||
XM_011528428.1 | Intron | XP_011526730.1 | ||||
XM_011528429.2 | Intron | XP_011526731.1 | ||||
XM_011528431.2 | Intron | XP_011526733.1 | ||||
XM_011528432.1 | Intron | XP_011526734.1 |