Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTTAGGCAGTGAACAATAGCT[A/G]TTGCCAGTTTGGAGCAATTTCCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 308385 MIM: 430000 | ||||||||||||||||||||
Literature Links: |
IL3RA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL3RA - interleukin 3 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267713.1 | 587 | Intron | NP_001254642.1 | |||
NM_002183.3 | 587 | Missense Mutation | TAT,TGT | Y,C 67 | NP_002174.1 | |
XM_005274431.4 | 587 | Missense Mutation | TAT,TGT | Y,C 67 | XP_005274488.1 | |
XM_005274432.1 | 587 | Missense Mutation | TAT,TGT | Y,C 67 | XP_005274489.1 | |
XM_017029491.1 | 587 | Missense Mutation | TAT,TGT | Y,C 67 | XP_016884980.1 |