Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAATCTGCCACAAACCAGAATGG[C/T]AGTAGAGCCAGTAACTTGCTGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRG3 - neuregulin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010848.3 | 43 | Intron | NP_001010848.2 | |||
NM_001165972.1 | 43 | Intron | NP_001159444.1 | |||
NM_001165973.1 | 43 | UTR 5 | NP_001159445.1 | |||
XM_005269444.4 | 43 | Intron | XP_005269501.1 | |||
XM_011539172.2 | 43 | Intron | XP_011537474.1 | |||
XM_011539173.2 | 43 | Intron | XP_011537475.1 | |||
XM_011539175.2 | 43 | Intron | XP_011537477.1 | |||
XM_011539178.2 | 43 | Intron | XP_011537480.1 | |||
XM_017015573.1 | 43 | Intron | XP_016871062.1 | |||
XM_017015574.1 | 43 | Intron | XP_016871063.1 | |||
XM_017015575.1 | 43 | Intron | XP_016871064.1 | |||
XM_017015576.1 | 43 | Intron | XP_016871065.1 | |||
XM_017015577.1 | 43 | Intron | XP_016871066.1 | |||
XM_017015578.1 | 43 | Intron | XP_016871067.1 | |||
XM_017015579.1 | 43 | Intron | XP_016871068.1 | |||
XM_017015580.1 | 43 | Intron | XP_016871069.1 | |||
XM_017015581.1 | 43 | Intron | XP_016871070.1 | |||
XM_017015582.1 | 43 | Intron | XP_016871071.1 | |||
XM_017015583.1 | 43 | Intron | XP_016871072.1 | |||
XM_017015584.1 | 43 | Intron | XP_016871073.1 |