Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGAGAATTCATCTTGTAGAAT[A/G]AAGTGAGAGCATGCTTGCGACACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH18A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH18A1 - aldehyde dehydrogenase 18 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017423.1 | Intron | NP_001017423.1 | ||||
NM_001323412.1 | Intron | NP_001310341.1 | ||||
NM_001323413.1 | Intron | NP_001310342.1 | ||||
NM_001323414.1 | Intron | NP_001310343.1 | ||||
NM_001323415.1 | Intron | NP_001310344.1 | ||||
NM_001323416.1 | Intron | NP_001310345.1 | ||||
NM_001323417.1 | Intron | NP_001310346.1 | ||||
NM_001323418.1 | Intron | NP_001310347.1 | ||||
NM_001323419.1 | Intron | NP_001310348.1 | ||||
NM_002860.3 | Intron | NP_002851.2 |