Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACGATATTGTCTGCGCTATTGC[C/T]CCAGCAACACCACCACAAAGTAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 139080 | ||||||||||||||||||||
Literature Links: |
SLC25A16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A16 - solute carrier family 25 member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324312.1 | 901 | Silent Mutation | GGA,GGG | G,G 251 | NP_001311241.1 | |
NM_001324313.1 | 901 | Silent Mutation | GGA,GGG | G,G 251 | NP_001311242.1 | |
NM_001324314.1 | 901 | Silent Mutation | GGA,GGG | G,G 153 | NP_001311243.1 | |
NM_001324315.1 | 901 | Silent Mutation | GGA,GGG | G,G 153 | NP_001311244.1 | |
NM_001324317.1 | 901 | Silent Mutation | GGA,GGG | G,G 117 | NP_001311246.1 | |
NM_152707.3 | 901 | Silent Mutation | GGA,GGG | G,G 251 | NP_689920.1 | |
XM_011540211.2 | 901 | Silent Mutation | GGA,GGG | G,G 251 | XP_011538513.1 | |
XM_017016695.1 | 901 | Intron | XP_016872184.1 |