Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCGCAGGGCACTGTCCTCGGG[C/G]GTACAGACCGTGGTCCCACATTCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM171A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM171A1 - family with sequence similarity 171 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010924.1 | 2349 | Silent Mutation | ACC,ACG | T,T 807 | NP_001010924.1 | |
XM_011519378.2 | 2349 | Silent Mutation | ACC,ACG | T,T 794 | XP_011517680.1 | |
XM_017015904.1 | 2349 | Silent Mutation | ACC,ACG | T,T 778 | XP_016871393.1 | |
XM_017015905.1 | 2349 | Silent Mutation | ACC,ACG | T,T 449 | XP_016871394.1 |